Magnesium-8-anilinonaphthalin-1-sulfonat Hydrat ≥95% (durch Perchlorsäure-Titration) für Fluoreszenz, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung


M-Clarity™ Qualitätsstufe = MQ200

A5144-25G A5144-5G
A5144-25GEA 353 EUR
A5144-25G A5144-5G
Magnesium-8-anilinonaphthalin-1-sulfonat Hydrat ≥95% (durch Perchlorsäure-Titration) für Fluoreszenz, Sigma-Aldrich®
Magnesium-8-anilinonaphthalin-1-sulfonat Hydrat

ANSA (8-Anilino-1-naphthalenesulfonic acid) is a hydrophobic polarity sensitive fluorescent dye useful as a site probe to detect conformational changes in cell and micelle membranes and molecules such as proteins. ANSA is also used as a fluorescent probe for the estimation of the critical micelle concentration (CMC) of surfactants.

Formel: C₃₂H₂₄MgN₂O₆S₂·xH₂O
Lagertemperatur: Room Temperature
MDL: MFCD16661224
CAS-Nummer: 18108-68-4
EINECS: 242-004-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c79f5f9067a9e9298716cd64e48c46c4ae277db8&vid=d491c40f96b503f8358720870192104ea5b66a1b&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR