D(+)-Raffinose Pentahydrat

Lieferant: PanReac AppliChem

Synonyme: Melitose Pentahydrat, Melitriose Pentahydrat, Gossypose Pentahydrat

A6882.0100
A6882.0100EA 189 EUR
A6882.0100
D(+)-Raffinose Pentahydrat
D(+)-Raffinose Pentahydrat
Formel: C₁₈H₃₂O₁₆·5H₂O
Molecular Weight: 594.52 g/mol
Schmelzpunkt: 80
Dichte: 1.516 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00071590
CAS-Nummer: 17629-30-0
EINECS: 208-146-9
Merck Index: 12,08279

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR