AppliClear water
Lieferant: PanReac AppliChem
A7004.0250
A7004.0250EA 70 EUR
A7004.0250
AppliClear water
Reinigungsmittel Desinfektionsmittel
Biocide is a quaternary ammonium compound.
  • Concentrate for the prevention of growth of algae and moulds in water baths
  • Non toxic
  • With colour indicator (blue to colourless)
  • 1 ml sufficient for 1 L
  • Germ-killing effect on microorganisms
AppliClear-water is fully active, when the colour indicator is blue. When colour turns to colourless, then refill with AppliClear-water.
Bitte beachten Biozidprodukte vorsichtig verwenden. Vor Gebrauch stets Etikett und Produktinformationen lesen.