Folsäure

Lieferant: PanReac AppliChem

Synonyme: Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9

A7092.0100 A7092.0025
APPCA7092.0100EA 257 EUR
APPCA7092.0100 A7092.0025
Folsäure
Folsäure
Formel: C₁₉H₁₉N₇O₆
Molecular Weight: 441.41 g/mol
MDL: MFCD00079305
CAS-Nummer: 59-30-3
EINECS: 200-419-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR