IPTG (Isopropyl-β-D-thiogalactopyranosid) (aus Galaktose aus Pflanzen) ≥99,5% (durch HPLC) dioxanfrei

Lieferant: PanReac AppliChem

Synonyme: IPTG

A7211.0025 A7211.1000
A7211.0025EA 683 EUR
A7211.0025 A7211.1000
IPTG (Isopropyl-β-D-thiogalactopyranosid) (aus Galaktose aus Pflanzen) ≥99,5% (durch HPLC) dioxanfrei
IPTG (Isopropyl-β-D-thiogalactopyranosid)

Isopropyl β-D-thiogalactopyranoside (IPTG) is derived from plant origin galactose.


  • Assay: min. 99,5%
  • Solid form
  • Dioxane free
  • Soluble in 5% water
  • Clear and colourless
  • Storage temperature: −20 °C


Assay (HPLC): min. 99.5 %
Solubility (5 %; H2O): clear, colorless
α20°C/D; 1 %, H2O: -29° - -33°
Dioxane: not detectable
Water (K.F.): max. 1.0 %


Bitte beachten Storage temperature: 2-8 °C

Formel: C₉H₁₈O₅S
Molecular Weight: 238.31 g/mol
Siedepunkt: 402
Lagertemperatur: Kühlschrank
MDL: MFCD00063273
CAS-Nummer: 367-93-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR