8-Aminopyrene-1,3,6-trisulphonic acid trisodium salt, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
Synonyme:
APTS
A7222-10MG
A7222-10MGEA
444
EUR
A7222-10MG
8-Aminopyrene-1,3,6-trisulphonic acid trisodium salt, Sigma-Aldrich®
8-Aminopyrene-1,3,6-trisulphonic acid trisodium salt
Fluorescent label for glycoproteins and saccharides.
8-Aminopyrene-1,3,6-trisulfonic acid trisodium salt (APTS) has been used for the detection of human milk oligosaccharides using multiplexed CGE with LIF (capillary gel electrophoresis with laser-induced
Formel:
C₁₆H₈NNa₃O₉S₃ Schmelzpunkt: ≥250 °C (Lit.) Lagertemperatur: Kühlschrank |
MDL:
MFCD00674494 CAS-Nummer: 196504-57-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...