Ammoniumheptamolybdat Tetrahydrat 81.0-83.0% (MoO₃-Basis) ACS, Fluka™

Lieferant: Honeywell Chemicals

A7302-100G A7302-1KG A7302-500G A7302-6X500G A7302-25KG
A7302-100GEA 77.28 EUR
A7302-100G A7302-1KG A7302-500G A7302-6X500G HONCA7302-25KG
Ammoniumheptamolybdat Tetrahydrat 81.0-83.0% (MoO₃-Basis) ACS, Fluka™
Ammoniumheptamolybdat Tetrahydrat
Formel: (NH₄)₆Mo₇O₂₄·4H₂O
Dichte: 2,498 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00167059
CAS-Nummer: 12054-85-2
EINECS: 234-320-9
Merck Index: 13,00536

Bestellen

Spezifikation Testergebnisse

Assay (as MoO₃) 81.0 - 83.0 %
Water insoluble matter Max. 0.005 %
Potassium (K) Max. 0.01 %
Magnesium (Mg) Max. 0.005 %
Sodium (Na) Max. 0.01 %
Heavy metals (ICP) ACS Max. 0.001 %
Chloride (Cl) Max. 0.002 %
Nitrate (NO₃) Max. 0.003 %
Phosphate, Arsenate, Silicate (as SiO₂) Max. 0.001 %
Phosphate (PO₄) Max. 5 ppm
Sulfate (SO₄) Max. 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR