APC-Cy5.5 Tandem 4 mg/ml, Suspension, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY


M-Clarity™ Qualitätsstufe = MQ200

A7472-.5ML
A7472-.5MLEA 495 EUR
A7472-.5ML
APC-Cy5.5 Tandem 4 mg/ml, Suspension, Sigma-Aldrich®
APC-Cy5.5 Tandem

Phycobiliprotein exhibiting exceptional absorbance and fluorescence characteristics due to a lack of susceptibility to internal and external fluorescence quenching. Ideal for highly sensitive studies in flow cytometry and immunoassays.


Allophycocyanin is a pigment which allows the energy trapped by phycobiliproteins to be transferred to the chloroplast lamellae. Physical form: 4 mg/ml solution in 60% saturated ammonium sulfate and 50 mM potassium phosphate.

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Blue
Appearance (Form) in 60% saturated Ammonium Sulfate and 50 mM Potassium Phosphate, pH 7.0. Suspension
Concentration Conforms 4 mg/ml
Fluorescence, Emission Max 655 - 665 nm
Relative Quantum Yield ≥1.8
Wavelength, Lambda Max 645 - 655 nm
UV Absorbance Ratio, A650/A280 ≥4.00
UV Absorbance Ratio, A650/A615 ≥1.30


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR