Cytochalasin B

Lieferant: PanReac AppliChem

A7657.0005
A7657.0005EA 243 EUR
A7657.0005
Cytochalasin B
Cytochalasin B
Formel: C₂₉H₃₇NO₅
Schmelzpunkt: 218…221 °C
MDL: MFCD00077704
CAS-Nummer: 14930-96-2
EINECS: 239-000-2
UN: 1544
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR