Acridine Orange 10-nonyl bromide (NAO), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: 3,6-Bis(dimethylamino)-10-nonylacridinium bromide, Nonyl acridine orange, NAO, 10-Nonylacridine orange bromide

MQ100

A7847-100MG
A7847-100MGEA 221 EUR
A7847-100MG
Acridine Orange 10-nonyl bromide (NAO), Sigma-Aldrich®
Acridine Orange 10-nonyl bromide (NAO)

Metachromatic dye useful as a mitochondrial probe in living cells. This dye has been used to analyse mitochondria by flow cytometry, characterise multidrug resistance, and measure changes in mitochondrial mass during apoptosis in rat thymocytes.

Formel: C₂₆H₃₈BrN₃
Molecular Weight: 472.50 g/mol
Schmelzpunkt: ≥250 °C (Lit.)
Lagertemperatur: Room Temperature
MDL: MFCD00467605
CAS-Nummer: 75168-11-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=728d283583601b02f9834f4f014e16c256a491ff&vid=728d283583601b02f9834f4f014e16c256a491ff&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) Orange to Red
Appearance (Form) Powder or Solid
Proton NMR Spectrum Conforms to Structure
EmM at lambda max 491 to 497 nm in Methanol 82 - 94
Wavelength 491 - 497 nm
Fluorescence emission max 515 - 523 nm
Purity (HPLC) ≥90%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR