Acridine orange (chlorid salt) -Lösung 10 mg/mL in Wasser, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: AO, C.I. 46005, 3,6-Bis(dimethylamino)acridine hydrochloride, Basic Orange 14, N,N,N’,N’-Tetramethyl-3,6-acridinediamine hydrochloride, 3,6-Acridinediamine, Acridine orange hydrochloride

A8097-10ML
A8097-10MLEA 145 EUR
A8097-10ML
Acridine orange (chlorid salt) -Lösung 10 mg/mL in Wasser, Sigma-Aldrich®
Acridinorange (Chloridsalz)

Acridine Orange hydrochloride solution has been used to study autophagic cell death. It has also been used for the staining of chromosomes.


  • hematology: suitable
  • histology: suitable


Acridine Orange is a metachromatic dye which can stain DNA, RNA and acid glycosaminoglycans. At low concentration it intercalates into DNA and precipitates RNA. However, at high concentration it denatures and precipitates both RNA and DNA. Acridine Orange is also used to analyze autophagy. It goes into acidic organelles in a pH-dependent manner. At neutral pH, acridine orange gives a green fluorescence and in acidic conditions, it accumulates in the acidic organelle giving a bright red fluorescence.

Formel: C₁₇H₁₉N₃·HCl·xH₂O
Molecular Weight: 301.81 g/mol (anhy)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012660
CAS-Nummer: 65-61-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR