Acriflavin fluorescent labeling reagent, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
|
Achtung
|
Synonyme:
3,6-Diamino-10-methylacridinium chloride, 3,6-Diamino-10-methylacridinium chloride-3,6-acridinediamine mix, Acriflavinium chloride 3,6-Acridinediamine mix, 3,6-Diamino-10-methylacridinium chloride mixt. with 3,6-diaminoacridine (proflavine), Euflavine, Trypaflavine Neutral, Euflavine, Acriflavine base
MQ200
Acriflavine also interacts with HIF-1α (hypoxia-inducible factor 1 α) and HIF-2α, and thereby suppresses dimer formation of HIF-1 and transcriptional function. It also has negative effect on tumor growth and vascularization. Acriflavine also exhibits trypanocidal, antibacterial and antiviral activities.
- Acriflavine is used to fluorescently label high molecular weight RNA molecules
- Excitation and emission wavelengths in various solvents:
- Methanol: λex = 424 nm; λem = 518 nm
- Ethanol: λex = 426 nm; λem = 524 nm
- Propanol: λex = 430 nm; λem = 512 nm
- Butanol: λex = 430 nm; λem = 526 nm
- Formamide: λex = 434 nm; λem = 524 nm
- Glycerol: λex = 432 nm; λem = 540 nm
- Water: λex = 416 nm; λem = 514 nm
Insoluble in ether, chloroform, and fixed oils. Utilised in fluorescence steady state measurements as a donor molecule (when paired with rhodamine 6G as the acceptor) to function as a pH sensor.
Acriflavine has been used in the agglutination test to distinguish between smooth and rough colony formation of Brucella melitensis.
It has been used as an AgO₂ (argonaute 2) inhibitor.
It has been used as an inhibitor of HIF-ARNT (hypoxia-inducible factor - aryl hydrocarbon receptor nuclear translocator) complex formation.
It has been used to study the bacteriocin production by Carnobacterium piscicola.
Formel:
C₂₇H₂₅ClN₆ Molecular Weight: 468.9806 g/mol Schmelzpunkt: 179...181 °C Lagertemperatur: Room Temperature |
MDL:
MFCD00064307 CAS-Nummer: 8048-52-0 |
|
Spezifikation Testergebnisse
Appearance (Form) | Powder |
Proton NMR spectrum | Conforms to Structure |
EmM, 462 nm (+/- 3 nm), C = 0.004 g/L in Methanol | ≥48000 |
Wavelength | 259 - 265 nm |
EmM, 262 nm (+/- 3 nm), C = 0.004 g/L in Methanol | ≥50000 |
Solubility (Colour) | Orange to Brown |
Solubility (Turbidity), 50 mg/mL in H₂O | Clear |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...