Acriflavin fluorescent labeling reagent, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: 3,6-Diamino-10-methylacridinium chloride, 3,6-Diamino-10-methylacridinium chloride-3,6-acridinediamine mix, Acriflavinium chloride 3,6-Acridinediamine mix, 3,6-Diamino-10-methylacridinium chloride mixt. with 3,6-diaminoacridine (proflavine), Euflavine, Trypaflavine Neutral, Euflavine, Acriflavine base

MQ200

A8126-100G A8126-25G
A8126-100GEA 354 EUR
A8126-100G A8126-25G
Acriflavin fluorescent labeling reagent, Sigma-Aldrich®
Acriflavin

Acriflavine also interacts with HIF-1α (hypoxia-inducible factor 1 α) and HIF-2α, and thereby suppresses dimer formation of HIF-1 and transcriptional function. It also has negative effect on tumor growth and vascularization. Acriflavine also exhibits trypanocidal, antibacterial and antiviral activities.


  • Acriflavine is used to fluorescently label high molecular weight RNA molecules
  • Excitation and emission wavelengths in various solvents:
  • Methanol: λex = 424 nm; λem = 518 nm
  • Ethanol: λex = 426 nm; λem = 524 nm
  • Propanol: λex = 430 nm; λem = 512 nm
  • Butanol: λex = 430 nm; λem = 526 nm
  • Formamide: λex = 434 nm; λem = 524 nm
  • Glycerol: λex = 432 nm; λem = 540 nm
  • Water: λex = 416 nm; λem = 514 nm


Insoluble in ether, chloroform, and fixed oils. Utilised in fluorescence steady state measurements as a donor molecule (when paired with rhodamine 6G as the acceptor) to function as a pH sensor.
Acriflavine has been used in the agglutination test to distinguish between smooth and rough colony formation of Brucella melitensis.
It has been used as an AgO₂ (argonaute 2) inhibitor.
It has been used as an inhibitor of HIF-ARNT (hypoxia-inducible factor - aryl hydrocarbon receptor nuclear translocator) complex formation.
It has been used to study the bacteriocin production by Carnobacterium piscicola.

Formel: C₂₇H₂₅ClN₆
Molecular Weight: 468.9806 g/mol
Schmelzpunkt: 179...181 °C
Lagertemperatur: Room Temperature
MDL: MFCD00064307
CAS-Nummer: 8048-52-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17cbd1fc8c21288f461a446d278a8c69e5c91322&vid=fc97931c6c6f70057c90a3bca4694e3a83670007&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Form) Powder
Proton NMR spectrum Conforms to Structure
EmM, 462 nm (+/- 3 nm), C = 0.004 g/L in Methanol ≥48000
Wavelength 259 - 265 nm
EmM, 262 nm (+/- 3 nm), C = 0.004 g/L in Methanol ≥50000
Solubility (Colour) Orange to Brown
Solubility (Turbidity), 50 mg/mL in H₂O Clear


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR