p-Amidinophenyl p-(6-amidino-2-indolyl)phenyl ether dihydrochloride, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
M-Clarity™ Qualitätsstufe = MQ200
A8675-5MG
A8675-5MGEA
927
EUR
A8675-5MG
p-Amidinophenyl p-(6-amidino-2-indolyl)phenyl ether dihydrochloride, Sigma-Aldrich®
p-Amidinophenyl p-(6-amidino-2-indolyl)phenyl ether dihydrochloride
This material has a structure similar to Granular Blue (CAS No. 71431-29-3). It has been referred to in some literature as Granular Blue, a fluorescent neuronal tracer.
Formel:
C₂₂H₁₉N₅O Molecular Weight: 442,34 g/mol Lagertemperatur: Room Temperature |
MDL:
MFCD00078924 CAS-Nummer: 55453-00-4 |
Spezifikation Testergebnisse
Appearance (Colour) | Yellow-Brown to Brown |
Appearance (Form) | Powder |
Solubility (Colour) | Yellow to Orange |
Solubility (Turbidity) 1 mg plus 0.1 ml H₂O | Clear |
Carbon | 50.8 - 60.8 % |
Nitrogen | 13.5 - 17.0 % |
Dye Content, Based on Nitrogen content (approx 90%) | ≥85 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...