Buffer, PBS tablets
Lieferant: PanReac AppliChem
PBS Tablets for preparation of 200 ml phosphate buffered saline (PBS). Composition of 140 mM sodium chloride, 2,7 mM potassium chloride and 10 mM phosphate.
- Ready to use in minutes
- Autoclavable
- Stable for approx. 12 months if stored at 2 to 8 °C
Deposit one tablet in flask or beaker placed on a magnetic stirrer. Add deionised water to 50% of the final volume at room temperature. Stir for a few minutes. Adjust the water to the final volume, stir until full dissolution. Autoclave or filter sterilise through a 0,22 µm filter into a sterile flask or bottle.