Auramin O, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr

Synonyme: Pyoctanunum aureum, 4,4′-(Imidocarbonyl)bis(N,N-dimethylaniline) monohydrochloride, Pyoktanin Yellow, 4,4'-(Iminomethylene)bis(N,N-dimethylaniline) hydrochloride, Basic Yellow 2, Canary Yellow

M-Clarity™ Qualitätsstufe = MQ200

A9655-25G
A9655-25GEA 151 EUR
A9655-25G
Auramin O, Sigma-Aldrich®
Auramin O
Formel: C₁₇H₂₂ClN₃
Molecular Weight: 303,84 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012484
CAS-Nummer: 2465-27-2
EINECS: 219-567-2
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efca0332c846bfba3f5c084d2d22494f62573194&vid=2ca58daf552de6a5ece20bf033700d86cbfe7965&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR