iFluor® 488, Azid

Lieferant: AAT BIOQUEST
Achtung

1000
AATB1000EA 460 EUR
AATB1000
iFluor® 488, Azid
iFluor®

Although FITC is still the most popular fluorescent labeling dye for preparing green fluorescent bioconjugates, there are certain limitations with FITC, such as severe photobleaching for microscope imaging and pH-sensitive fluorescence.


  • pH-insensitive
  • Azide reactive
  • High photostability
  • Brighter signal

ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46c1983ca7b8c82915928198be83e812bef145de&vid=fceff12a6481d8b42a270191bc1e1b0adf827de9&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR