PBS Tablet (Phosphate Buffered Saline Tablet)
Lieferant: AAT BIOQUEST
10010
AATB10010EA
62.5
EUR
AATB10010
PBS Tablet (Phosphate Buffered Saline Tablet)
Puffer
Elektrophorese-Puffer
Waschpuffer
Each PBS tablet prepares 100 mL of a 1X PBS solution containing 137 mM sodium chloride, 10 mM phosphate buffer, and 2.7 mM potassium chloride (pH 7.4).
- Designed for ease of use
- Saves time
PBS (phosphate buffered saline) is an isotonic buffer that closely mimics the human body's pH, osmolarity, and ion concentrations.
Bitte beachten For research use only.