Stains-all ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
1-Ethyl-2-[3-(1-ethylnaphtho[1,2-d]thiazolin-2-ylidene)-2-methylpropenyl]naphtho[1,2-d]thiazolium bromide, 3,3′-Diethyl-9-methyl-4,5,4′,5′-dibenzothiacarbocyanine
10050
AATB10050EA
120
EUR
AATB10050
Stains-all ≥95%
Stains-all
Stains-all is suitable for differential staining of nucleic acids and proteins.
Formel:
C₃₀H₂₇BrN₂S₂ Molecular Weight: 559,584 g/mol |
MDL:
MFCD00078788 CAS-Nummer: 7423-31-6 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...