iFluor® 555 fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

1007
AATB1007EA 292 EUR
AATB1007
iFluor® 555 fluoreszierende Probe
iFluor®

The tetrazine-trans-cyclooctene (TCO) ligation constitutes a non-toxic biomolecule labeling method of unparalleled speed.


  • pH-insensitive
  • Azide reactive
  • High photostability
  • Brighter signal

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36e3b88f3435259dc66f9db5fdfb86b3f6b60506&vid=67e951a26610d66b038e5c1d5c0a068e0ae4e587&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR