Amplite® Colorimetric Malondialdehyde (MDA) Quantitation Kit
Lieferant: AAT BIOQUEST
10070
AATB10070EA
537
EUR
AATB10070
Amplite® Colorimetric Malondialdehyde (MDA) Quantitation Kit
Tests
Enzymtests
Malondialdehyde (MDA) is natural byproduct of lipid peroxidation and is widely used as a key indicator to determine the oxidative stress and free radical formation.
- In-house QC testing
- Sensitive
- Suggested protocol available
Bitte beachten For research use only.