iFluor® 555 fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
1016
AATB1016EA
292
EUR
AATB1016
iFluor® 555 fluoreszierende Probe
iFluor®
The tetrazine-trans-cyclooctene (TCO) ligation constitutes a non-toxic biomolecule labeling method of unparalleled speed.
- pH-insensitive
- Azide reactive
- High photostability
- Brighter signal
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...