iFluor® 570 succinimidyl ester
Lieferant: AAT BIOQUEST
1034
AATB1034EA 309 EUR
AATB1034
iFluor® 570 succinimidyl ester
iFluor® 570 succinimidyl ester
iFluor® 570 family has fluorescence that is pH-independent from pH 3 to 11.
  • Stable for 12 months
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
Bitte beachten For Research Use Only