iFluor® 690 succinimidyl ester
Lieferant: AAT BIOQUEST
1042
AATB1042EA 429 EUR
AATB1042
iFluor® 690 succinimidyl ester
iFluor® 690 succinimidyl ester
iFluor® 690 dyes have fluorescence excitation and emission maxima of ~685 nm and ~704 nm respectively.
  • Stable for 12 months
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, particularly antibodies.
Bitte beachten For Research Use Only