iFluor® 675 succinimidyl ester
Lieferant: AAT BIOQUEST
1043
AATB1043EA
429
EUR
AATB1043
iFluor® 675 succinimidyl ester
iFluor® 675 succinimidyl ester
iFluor® 675 dyes have fluorescence excitation and emission maxima of ~677 nm and ~710 nm respectively.
- Stable for 12 months
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
Bitte beachten For Research Use Only