iFluor® 500 succinimidyl ester
Lieferant: AAT BIOQUEST
1044
AATB1044EA 420 EUR
AATB1044
iFluor® 500 succinimidyl ester
iFluor® 500 succinimidyl ester
iFluor® 500 dyes have fluorescence excitation and emission maxima of ~500 nm and ~520 nm respectively.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only