iFluor® 597 succinimidyl ester
Lieferant: AAT BIOQUEST
1050
AATB1050EA
429
EUR
AATB1050
iFluor® 597 succinimidyl ester
iFluor® 597 succinimidyl ester
iFluor® 597 is a unique color for fluorescence imaging and flow cytometry applications.
- Stable for 12 months
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, particularly antibodies.
Bitte beachten For Research Use Only