iFluor® 620 succinimidyl ester
Lieferant: AAT BIOQUEST
1051
AATB1051EA 429 EUR
AATB1051
iFluor® 620 succinimidyl ester
iFluor® 620 succinimidyl ester
iFluor® 620 dyes have fluorescence excitation and emission maxima of ~621 nm and ~636 nm respectively.
  • Stable for 12 months
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
Bitte beachten For Research Use Only