iFluor® 405, Maleimide
Lieferant: AAT BIOQUEST
Achtung
1053
AATB1053EA 307 EUR
AATB1053
iFluor® 405, Maleimide
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=632676356b59228777e61eb6a3c8765809ce804d&vid=719fdda64f60369bebaf7627f70c1e70e192fe0a&prid=web&clid=SAPDAM