iFluor® 568, Maleimide
Lieferant: AAT BIOQUEST
Achtung
1055
AATB1055EA 307 EUR
AATB1055
iFluor® 568, Maleimide
iFluor®
iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7624d9b2a9515e5ba638425ae234e1cb803cc6dd&vid=2e43bf6d618445ff6269c3dba3b97cece9a7d5c2&prid=web&clid=SAPDAM