iFluor® 568, Maleimide
Lieferant: AAT BIOQUEST
|
Achtung
|
1055
AATB1055EA
307
EUR
AATB1055
iFluor® 568, Maleimide
iFluor®
iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
- Bright and photostable
- Minimal quenching
- Stable conjugation
ADR:
5,II |