iFluor® 633, Maleimide
Lieferant: AAT Bioquest
Achtung
1056
AATB1056EA 307 EUR
AATB1056
iFluor® 633, Maleimide
iFluor®
AAT Bioquest\'s iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5617a08f5770ef929c7d1a6dc3ba8197a2c6be56&vid=538474aa78f083f0ccc1db6379122faae20a32b3&prid=web&clid=SAPDAM