iFluor® 450, Maleimide

Lieferant: AAT BIOQUEST
Achtung

1057
AATB1057EA 307 EUR
AATB1057
iFluor® 450, Maleimide
iFluor®

iFluor® dyes are optimized for labeling proteins, in particular, antibodies.


  • pH-insensitive
  • Azide reactive
  • High photostability
  • Brighter signal

ADR: 5,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR