iFluor® 546 maleimide
Lieferant: AAT BIOQUEST
1059
AATB1059EA 309 EUR
AATB1059
iFluor® 546 maleimide
iFluor® 546 maleimide
iFluor® 546 dye might be the brightest fluorophore available that is well excited by the 546 nm laser or LED light sources.
  • Stable for 12 months
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, particularly antibodies.
Bitte beachten For Research Use Only