iFluor® 350, Maleimide
Lieferant: AAT BIOQUEST
Achtung
1060
AATB1060EA 307 EUR
AATB1060
iFluor® 350, Maleimide
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
ADR: 5,II