iFluor® 350, Maleimide
Lieferant: AAT BIOQUEST
|
Achtung
|
1060
AATB1060EA
307
EUR
AATB1060
iFluor® 350, Maleimide
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
- Bright and photostable
- Minimal quenching
- Stable conjugation
ADR:
5,II |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)