iFluor® 532, Maleimide
Lieferant: AAT BIOQUEST
|
Achtung
|
1061
AATB1061EA
307
EUR
AATB1061
iFluor® 532, Maleimide
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
- pH-insensitive
- Azide reactive
- High photostability
- Brighter signal
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...