iFluor® 647, Maleimide

Lieferant: AAT BIOQUEST
Achtung

1065
AATB1065EA 307 EUR
AATB1065
iFluor® 647, Maleimide
iFluor®

AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.


  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
  • pH independent fluorescence between 3-11

ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR