iFluor® 700, Maleimide
Lieferant: AAT BIOQUEST
Achtung
1067
AATB1067EA 307 EUR
AATB1067
iFluor® 700, Maleimide
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=192560a74f9587cecc21d1b94a59c410db0a2f0c&vid=f925e1da29e8546489836d6198e6385a47575941&prid=web&clid=SAPDAM