iFluor® 405, Amin
Lieferant: AAT BIOQUEST
Achtung
1071
AATB1071EA 307 EUR
AATB1071
iFluor® 405, Amin
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e1df5135ab3082ab159d68da5c47c3c1c0e0151&vid=9301ae4c4bcda426682c68a5d2aea2355074a1f2&prid=web&clid=SAPDAM