iFluor® 488, Amin

Lieferant: AAT BIOQUEST
Achtung

1072
AATB1072EA 307 EUR
AATB1072
iFluor® 488, Amin
iFluor®

Although FITC is still the most popular fluorescent labeling dye for preparing green fluorescent bioconjugates, there are certain limitations with FITC, such as severe photobleaching for microscope imaging and pH-sensitive fluorescence.


  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
  • pH independent fluorescence between 4-10

ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=64a62f4fb74e7736349752c6612ca131bfff50a6&vid=4976834f6543e9f7cf96584b5b53a973024d822e&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR