iFluor® 647, Amin
Lieferant: AAT BIOQUEST
Achtung
1074
AATB1074EA 307 EUR
AATB1074
iFluor® 647, Amin
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d94be67ab55fb2774e79dfa75dace2a7efcf79c&vid=163b5ff1bc38e66090a4ef0bcf7677adcad15b4f&prid=web&clid=SAPDAM