iFluor® 660, Amin
Lieferant: AAT BIOQUEST
Achtung
1075
AATB1075EA 307 EUR
AATB1075
iFluor® 660, Amin
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c50d991d2abefc16c19041aa960b1a0413b20a09&vid=38f5e7fe7cb6353e654bb8a5d3d13d72ddfd3ed7&prid=web&clid=SAPDAM