iFluor® 750, Amin
Lieferant: AAT BIOQUEST
|
Achtung
|
1079
AATB1079EA
307
EUR
AATB1079
iFluor® 750, Amin
iFluor®
iFluor® dyes are developed for labeling proteins, in particular, antibodies.
- Bright and photostable
- Minimal quenching
- Stable conjugation
ADR:
5,II |