iFluor® 555, Hydrazid
Lieferant: AAT BIOQUEST
|
Achtung
|
1083
AATB1083EA
307
EUR
AATB1083
iFluor® 555, Hydrazid
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
- Bright and photostable
- Minimal quenching
- Stable conjugation
- pH independent fluorescence between 3-11
ADR:
5,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...