iFluor® 647, Alkin
Lieferant: AAT BIOQUEST
Achtung
1090
AATB1090EA 307 EUR
AATB1090
iFluor® 647, Alkin
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf1a9731f9d6c7e00868249e63fe39a146490f64&vid=48c777ee77931432ac577e27eef53464f54698c6&prid=web&clid=SAPDAM