iFluor® 555, Alkin
Lieferant: AAT BIOQUEST
Achtung
1092
AATB1092EA 307 EUR
AATB1092
iFluor® 555, Alkin
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright and photostable
  • Minimal quenching
  • Stable conjugation
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e4d2a0db57c6bf97d408d9d82d8b268882060e9&vid=e2f9a8bf8913d8b515964a7defaa8a5f1e3a1f4d&prid=web&clid=SAPDAM