iFluor® 555, Alkin
Lieferant: AAT BIOQUEST
|
Achtung
|
1092
AATB1092EA
307
EUR
AATB1092
iFluor® 555, Alkin
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
- Bright and photostable
- Minimal quenching
- Stable conjugation
ADR:
5,II |