iFluor® 555, Azid
Lieferant: AAT BIOQUEST
|
Achtung
|
1093
AATB1093EA
307
EUR
AATB1093
iFluor® 555, Azid
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
- Bright and photostable
- Minimal quenching
- Stable conjugation
ADR:
5 II |