iFluor® 405 azide
Lieferant: AAT BIOQUEST
1094
AATB1094EA
309
EUR
AATB1094
iFluor® 405 azide
iFluor® 405 azide
iFluor® 405 azide is reasonably stable and shows good reactivity and selectivity with the alkyne group in click chemistry.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only