ReadiLink™ Rapid mFluor™ Violet 450 Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
Lieferant: AAT BIOQUEST
1100
AATB1100EA 230 EUR
AATB1100
ReadiLink™ Rapid mFluor™ Violet 450 Antibody Labelling Kit 'Microscale Optimised for Labelling 50 µg Antibody Per Reaction' (2 Labellings)
Proteinkonjugate und Markierungen
AAT Bioquest's mFluor™ dyes are developed for flow cytometry-focused applications.
  • Suitable for Pacific Blue™ channel
  • robust and simple antibody labelling protocol
  • no column purification required
Bitte beachten For research use only.