DIG-HRP Conjugate (1 mg)
Lieferant: AAT BIOQUEST
11023
AATB11023EA
218
EUR
AATB11023
DIG-HRP Conjugate (1 mg)
Enzyme
Digoxigenin (DIG)-conjugated horse radish peroxidase (HRP) is an excellent tool for biological detections with ELISA and other DIG-coupled assays.
- Selective binding
- ELISA development applications
Bitte beachten For research use only.