ReadiUse™ Preactivated HRP NHS Ester
Lieferant: AAT BIOQUEST
11025
AATB11025EA 152 EUR
AATB11025
ReadiUse™ Preactivated HRP NHS Ester
Tests Immunologische Tests
ReadiUse™ Preactivated HRP NHS ester is the mono NHS ester of HRP.
  • Ready-to-use
  • Highly useful antibody label
  • Widely used for immunoassays
  • Multiple applications
Bitte beachten For research use only.